siRNA Details
| Gene Symbol: Slc12a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020333.2 | NP_065066.2 | 20 | 3063 | ||
| GenBank mRNA | AF332063.1 | - | 20 | 2733 | ||
| AF332064.1 | - | 20 | 2733 | |||
| AK122460.1 | - | 20 | 2797 | |||
| AK147262.1 | - | 20 | 3065 | |||
| BC054808.1 | - | 20 | 2729 | 6838880 | ||
| BC057624.1 | - | 1 | 113 | |||
| KJ535320.1 | - | 20 | 2754 | |||
| KJ535321.1 | - | 20 | 2754 | |||
| KJ535322.1 | - | 20 | 2910 | |||

Slc12a5
Genomic Map