siRNA Details
| Gene Symbol: Slc12a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020333.2 | NP_065066.2 | 9 | 1625 | ||
| GenBank mRNA | AF332063.1 | - | 9 | 1295 | ||
| AF332064.1 | - | 9 | 1295 | |||
| AK122460.1 | - | 9 | 1359 | |||
| AK147262.1 | - | 9 | 1627 | |||
| BC054808.1 | - | 9 | 1291 | 6838880 | ||
| KJ535320.1 | - | 9 | 1316 | |||
| KJ535321.1 | - | 9 | 1316 | |||
| KJ535322.1 | - | 9 | 1472 | |||

Slc12a5
Genomic Map