siRNA Details
| Gene Symbol: Dolpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290508.1 | NP_001277437.1 | 3 | 514 | ||
| NM_001290509.1 | NP_001277438.1 | 2 | 307 | |||
| NM_020329.4 | NP_065062.1 | 2 | 307 | |||
| GenBank mRNA | AB030189.1 | - | 2 | 147 | ||
| AK002544.1 | - | 3 | 353 | |||
| AK044141.1 | - | 2 | 146 | |||
| AK053923.1 | - | 2 | 136 | |||
| AK088065.1 | - | 2 | 151 | |||
| BC026544.1 | - | 2 | 131 | 4975127 | ||
| BY133699.1 | - | 2 | 232 | |||

Dolpp1
Genomic Map