siRNA Details
| Gene Symbol: Tbx20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205085.1 | NP_001192014.1 | 5 | 1156 | ||
| NM_020496.3 | NP_065242.1 | 5 | 1156 | |||
| NM_194263.2 | NP_919239.1 | 5 | 1156 | |||
| GenBank mRNA | AF260557.1 | - | 5 | 1161 | ||
| AF306667.1 | - | 5 | 1094 | |||
| AJ277486.1 | - | 5 | 698 | |||
| AK052155.1 | - | 5 | 1138 | |||
| AK052389.1 | - | 5 | 1139 | |||
| AK141251.1 | - | 5 | 926 | |||
| BY385297.1 | - | 3 | 187 | |||

Tbx20
Genomic Map