siRNA Details
| Gene Symbol: Ly6i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020498.2 | NP_065244.2 | 3 | 633 | ||
| GenBank mRNA | AF232024.1 | - | 3 | 555 | ||
| BC125350.1 | - | 3 | 633 | 40129865 | ||
| BC132052.1 | - | 3 | 633 | 40130329 | ||
| BC145087.1 | - | 4 | 375 | 9053635 | ||

Ly6i
Genomic Map