siRNA Details
| Gene Symbol: Olfr653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147074.2 | NP_667285.2 | 1 | 638 | ||
| GenBank mRNA | BC116962.1 | - | 1 | 638 | 40126281 | |
| BC120700.1 | - | 1 | 638 | 40129623 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr653| Gene Symbol: Olfr653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147074.2 | NP_667285.2 | 1 | 638 | ||
| GenBank mRNA | BC116962.1 | - | 1 | 638 | 40126281 | |
| BC120700.1 | - | 1 | 638 | 40129623 | ||