siRNA Details
Gene Symbol: Tas2r108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020502.1 | NP_065248.1 | 1 | 205 | ||
GenBank mRNA | AY916507.1 | - | 1 | 205 | ||
BC116952.1 | - | 1 | 268 | 40126271 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tas2r108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020502.1 | NP_065248.1 | 1 | 205 | ||
GenBank mRNA | AY916507.1 | - | 1 | 205 | ||
BC116952.1 | - | 1 | 268 | 40126271 |