siRNA Details
| Gene Symbol: Prss21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020487.4 | NP_065233.2 | 6 | 861 | ||
| GenBank mRNA | AB033822.1 | - | 6 | 847 | ||
| AB059415.1 | - | 6 | 859 | |||
| AF176209.2 | - | 6 | 861 | |||
| AK006271.1 | - | 6 | 833 | |||
| AY005145.1 | - | 6 | 838 | |||
| BC049588.1 | - | 6 | 875 | |||
| BC116755.1 | - | 6 | 859 | 40126074 | ||
| BC116757.1 | - | 6 | 859 | 40126076 | ||

Prss21
Genomic Map