siRNA Details
| Gene Symbol: Xpo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020506.1 | NP_065252.1 | 21 | 3129 | ||
| GenBank mRNA | AF145021.1 | - | 21 | 3129 | ||
| AK042783.1 | - | 4 | 471 | |||
| AK129430.1 | - | 21 | 3124 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xpo4| Gene Symbol: Xpo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020506.1 | NP_065252.1 | 21 | 3129 | ||
| GenBank mRNA | AF145021.1 | - | 21 | 3129 | ||
| AK042783.1 | - | 4 | 471 | |||
| AK129430.1 | - | 21 | 3124 | |||