siRNA Details
| Gene Symbol: Retnla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020509.3 | NP_065255.2 | 3 | 274 | ||
| GenBank mRNA | AF205951.1 | - | 2 | 207 | ||
| AF290872.1 | - | 3 | 312 | |||
| AF316397.2 | - | 3 | 216 | |||
| AF323082.1 | - | 3 | 284 | |||
| AK007553.1 | - | 3 | 219 | |||
| BC029248.1 | - | 2 | 214 | 4189285 | ||

Retnla
Genomic Map