siRNA Details
| Gene Symbol: Retn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204959.1 | NP_001191888.1 | 4 | 346 | ||
| NM_022984.4 | NP_075360.1 | 4 | 346 | |||
| GenBank mRNA | AF290870.1 | - | 4 | 339 | ||
| AF323080.1 | - | 4 | 345 | |||
| AK131613.1 | - | 4 | 347 | |||
| AK131869.1 | - | 4 | 343 | |||
| BC051196.1 | - | 4 | 332 | 1448672 | ||

Retn
Genomic Map