siRNA Details
| Gene Symbol: Cxcl14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019568.2 | NP_062514.2 | 4 | 1311 | ||
| GenBank mRNA | AB041614.1 | - | 4 | 1311 | ||
| AF144754.1 | - | 4 | 1135 | |||
| AF192557.1 | - | 4 | 1309 | |||
| AK004615.1 | - | 4 | 1156 | |||
| AK076112.1 | - | 4 | 1153 | |||
| BC079661.1 | - | 3 | 951 | |||
| BC147274.1 | - | 4 | 1055 | 9056321 | ||
| BC147275.1 | - | 4 | 1055 | 9056329 | ||

Cxcl14
Genomic Map