siRNA Details
Gene Symbol: Olfr140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020515.1 | NP_065261.1 | 1 | 235 | ||
GenBank mRNA | BC111111.1 | - | 1 | 232 | ||
BC141888.1 | - | 1 | 234 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020515.1 | NP_065261.1 | 1 | 235 | ||
GenBank mRNA | BC111111.1 | - | 1 | 232 | ||
BC141888.1 | - | 1 | 234 |