siRNA Details
| Gene Symbol: Bcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020486.2 | NP_065232.1 | 8 | 1094 | ||
| GenBank mRNA | AB035511.1 | - | 8 | 1055 | ||
| AF109160.1 | - | 8 | 1094 | |||
| AF221507.1 | - | 8 | 1044 | |||
| AF346663.1 | - | 8 | 1032 | |||
| AK040564.1 | - | 8 | 1078 | |||
| AK054001.1 | - | 8 | 1075 | |||
| AK075704.1 | - | 8 | 1059 | |||
| AK158439.1 | - | 8 | 1060 | |||
| BC004826.1 | - | 8 | 1078 | 2616148 | ||

Bcam
Genomic Map