siRNA Details
| Gene Symbol: Bcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020486.2 | NP_065232.1 | 6, 7 | 832 | ||
| GenBank mRNA | AB035511.1 | - | 6, 7 | 793 | ||
| AF109160.1 | - | 6, 7 | 832 | |||
| AF221507.1 | - | 6, 7 | 782 | |||
| AF346663.1 | - | 6, 7 | 770 | |||
| AK040564.1 | - | 6, 7 | 816 | |||
| AK054001.1 | - | 6, 7 | 813 | |||
| AK075704.1 | - | 6, 7 | 797 | |||
| AK158439.1 | - | 6, 7 | 795 | |||
| BC004826.1 | - | 6, 7 | 816 | 2616148 | ||

Bcam
Genomic Map