siRNA Details
| Gene Symbol: LCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005565.3 | NP_005556.1 | 4 | 451 | ||
| GenBank mRNA | AK292890.1 | - | 4 | 412 | ||
| AK310369.1 | - | 4 | 392 | |||
| BC016618.1 | - | 4 | 388 | 4476274 | ||
| BG911397.1 | - | 4 | 453 | |||
| BT007273.1 | - | 4 | 244 | |||
| FN432830.1 | - | 4 | 244 | |||
| U20158.1 | - | 4 | 499 | |||

LCP2
Genomic Map