siRNA Details
| Gene Symbol: Nelfcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020580.3 | NP_065605.2 | 11 | 1364 | ||
| GenBank mRNA | AJ238741.1 | - | 11 | 1298 | ||
| AJ238742.1 | - | 11 | 1364 | |||
| AJ238743.1 | - | 11 | 1318 | |||
| AK010360.1 | - | 11 | 1311 | |||
| AK049418.1 | - | 11 | 1336 | |||
| AK159866.1 | - | 11 | 1365 | |||
| AK178814.1 | - | 2 | 139 | |||
| BC006959.1 | - | 11 | 1325 | 3707929 | ||
| BC031747.1 | - | 11 | 1322 | 3673038 | ||

Nelfcd
Genomic Map