siRNA Details
| Gene Symbol: Wdr46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020603.2 | NP_065628.1 | 8 | 793 | ||
| GenBank mRNA | AK145068.1 | - | 8 | 848 | ||
| BC013527.1 | - | 8 | 836 | |||
| BC026431.1 | - | 5 | 417 | |||
| BC046977.1 | - | 8 | 793 | 6315172 | ||
| KJ650211.1 | - | 8 | 778 | |||

Wdr46
Genomic Map