siRNA Details
| Gene Symbol: Jph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020604.2 | NP_065629.1 | 4 | 1556 | ||
| GenBank mRNA | AB024445.1 | - | 4 | 1465 | ||
| BC120839.1 | - | 4 | 1556 | 40129762 | ||
| BC137669.1 | - | 4 | 1556 | 8860688 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jph1| Gene Symbol: Jph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020604.2 | NP_065629.1 | 4 | 1556 | ||
| GenBank mRNA | AB024445.1 | - | 4 | 1465 | ||
| BC120839.1 | - | 4 | 1556 | 40129762 | ||
| BC137669.1 | - | 4 | 1556 | 8860688 | ||