siRNA Details
| Gene Symbol: Cramp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020608.3 | NP_065633.2 | 18 | 3327 | ||
| GenBank mRNA | AK089393.1 | - | 18 | 3587 | ||
| AK129356.1 | - | 9 | 1503 | |||
| AK147411.1 | - | 18 | 3328 | |||
| AK147449.1 | - | 18 | 3328 | |||

Cramp1l
Genomic Map