siRNA Details
| Gene Symbol: Cramp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020608.3 | NP_065633.2 | 8 | 1068 | ||
| GenBank mRNA | AK147411.1 | - | 8 | 1069 | ||
| AK147449.1 | - | 8 | 1069 | |||
| AK193094.1 | - | 2 | 132 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cramp1l| Gene Symbol: Cramp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020608.3 | NP_065633.2 | 8 | 1068 | ||
| GenBank mRNA | AK147411.1 | - | 8 | 1069 | ||
| AK147449.1 | - | 8 | 1069 | |||
| AK193094.1 | - | 2 | 132 | |||