siRNA Details
| Gene Symbol: Zc3h8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020594.2 | NP_065619.2 | 2, 3 | 261 | ||
| GenBank mRNA | AF061961.1 | - | 2, 3 | 269 | ||
| AK021376.1 | - | 2, 3 | 265 | |||
| AK132679.1 | - | 2, 3 | 208 | |||
| AK164491.1 | - | 2, 3 | 208 | |||
| BC048687.1 | - | 2, 3 | 193 | 6334861 | ||
| CJ167668.1 | - | 2, 3 | 262 | |||

Zc3h8
Genomic Map