siRNA Details
| Gene Symbol: Plin4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020568.3 | NP_065593.2 | 6 | 3363 | ||
| GenBank mRNA | AF064748.1 | - | 6 | 3374 | ||
| AK173286.1 | - | 6 | 2095 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plin4| Gene Symbol: Plin4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020568.3 | NP_065593.2 | 6 | 3363 | ||
| GenBank mRNA | AF064748.1 | - | 6 | 3374 | ||
| AK173286.1 | - | 6 | 2095 | |||