siRNA Details
| Gene Symbol: Slc15a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021301.3 | NP_067276.2 | 14 | 1351 | ||
| GenBank mRNA | AF111811.1 | - | 14 | 1351 | ||
| AK128939.1 | - | 14 | 1184 | |||
| BC138627.1 | - | 14 | 1308 | 8861647 | ||
| BC138628.1 | - | 14 | 1308 | 8861648 | ||

Slc15a2
Genomic Map