siRNA Details
| Gene Symbol: Stk32c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162540.1 | NP_001156012.1 | 11 | 1141 | ||
| NM_021302.3 | NP_067277.2 | 11 | 1441 | |||
| GenBank mRNA | AB041542.1 | - | 11 | 1421 | ||
| AK046439.1 | - | 11 | 1419 | |||
| BC026457.1 | - | 11 | 1426 | 4481218 | ||

Stk32c
Genomic Map