siRNA Details
| Gene Symbol: Noc2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021303.2 | NP_067278.2 | 8 | 888 | ||
| GenBank mRNA | AK076287.1 | - | 7 | 930 | ||
| AK078560.1 | - | 8 | 864 | |||
| AK133834.1 | - | 8 | 888 | |||
| AK157765.1 | - | 8 | 881 | |||
| AK164071.1 | - | 2 | 2505 | |||
| AK184541.1 | - | 4 | 333 | |||

Noc2l
Genomic Map