siRNA Details
| Gene Symbol: Jmy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021310.3 | NP_067285.2 | 1 | 1429 | ||
| GenBank mRNA | AB221690.1 | - | 1 | 927 | ||
| AF201390.1 | - | 1 | 1053 | |||
| AK139934.1 | - | 1 | 1058 | |||
| AK141957.1 | - | 1 | 1430 | |||
| BC020052.1 | - | 1 | 1354 | |||
| BC069906.1 | - | 1 | 1283 | |||
| BC090835.1 | - | 1 | 1310 | 5705427 | ||

Jmy
Genomic Map