siRNA Details
| Gene Symbol: Rnf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF171060.1 | - | 6, 7 | 449 | ||
| AK002399.1 | - | 6, 7 | 470 | |||
| AK011171.1 | - | 3, 4 | 178 | |||
| AK019236.1 | - | 3, 4 | 176 | |||
| AK171752.1 | - | 6, 7 | 447 | |||
| BC022715.1 | - | 6, 7 | 435 | |||
| BC037100.1 | - | 6, 7 | 460 | |||
| BC048821.1 | - | 6, 7 | 457 | 3966642 | ||

Rnf25
Genomic Map