siRNA Details
| Gene Symbol: Tacc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004468.2 | NP_001004468.1 | 7 | 6697 | ||
| NM_021314.4 | NP_067289.2 | 3 | 1610 | |||
| NM_206856.3 | NP_996738.2 | 6 | 1576 | |||
| GenBank mRNA | AB041546.1 | - | 3 | 1611 | ||
| AK050985.1 | - | 4 | 1505 | |||
| AK145721.1 | - | 4 | 1518 | |||
| AK165194.1 | - | 3 | 1550 | |||
| AK165603.1 | - | 3 | 1401 | |||
| AY177410.1 | - | 5 | 1588 | |||
| BC004057.1 | - | 1 | 839 | |||

Tacc2
Genomic Map