siRNA Details
| Gene Symbol: Tacc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004468.2 | NP_001004468.1 | 7 | 6062 | ||
| NM_021314.4 | NP_067289.2 | 3 | 975 | |||
| NM_206856.3 | NP_996738.2 | 6 | 941 | |||
| GenBank mRNA | AB041546.1 | - | 3 | 976 | ||
| AK050985.1 | - | 4 | 870 | |||
| AK145721.1 | - | 4 | 883 | |||
| AK165194.1 | - | 3 | 915 | |||
| AK165603.1 | - | 3 | 766 | |||
| AY177410.1 | - | 5 | 953 | |||
| BC004057.1 | - | 1 | 204 | |||

Tacc2
Genomic Map