siRNA Details
| Gene Symbol: Tacc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004468.2 | NP_001004468.1 | 12, 13 | 7515 | ||
| NM_021314.4 | NP_067289.2 | 8, 9 | 2350 | |||
| NM_206856.3 | NP_996738.2 | 11, 12 | 2316 | |||
| GenBank mRNA | AB041546.1 | - | 8, 9 | 2351 | ||
| AK050985.1 | - | 8, 9 | 2209 | |||
| AK145721.1 | - | 8, 9 | 2210 | |||
| AK165194.1 | - | 7, 8 | 2254 | |||
| AK165603.1 | - | 7, 8 | 2105 | |||
| AY177410.1 | - | 10, 11 | 2328 | |||
| BC004057.1 | - | 5, 6 | 1621 | |||
| BI110807.1 | - | 2, 3 | 138 | |||

Tacc2
Genomic Map