siRNA Details
| Gene Symbol: Noc3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021315.2 | NP_067290.2 | 9, 10 | 1254 | ||
| GenBank mRNA | AB077991.1 | - | 9, 10 | 1217 | ||
| AK134051.1 | - | 9, 10 | 1212 | |||
| AK168944.1 | - | 9, 10 | 1255 | |||
| BC031132.1 | - | 9, 10 | 1217 | 4954951 | ||

Noc3l
Genomic Map