siRNA Details
| Gene Symbol: Tbx21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019507.2 | NP_062380.2 | 3 | 852 | ||
| GenBank mRNA | AF093099.1 | - | 3 | 810 | ||
| AF241242.1 | - | 3 | 842 | |||
| AK054495.1 | - | 3 | 851 | |||
| AK156271.1 | - | 3 | 853 | |||
| AY836551.1 | - | 2 | 165 | |||
| BC137986.1 | - | 3 | 770 | 8861005 | ||
| BC137988.1 | - | 3 | 770 | 8861007 | ||

Tbx21
Genomic Map