siRNA Details
| Gene Symbol: Rbak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045482.2 | NP_001038947.1 | 5 | 1885 | ||
| NM_021326.3 | NP_067301.1 | 5 | 1985 | |||
| GenBank mRNA | AF226870.1 | - | 5 | 1843 | ||
| AK051009.1 | - | 5 | 1986 | |||
| BC059913.1 | - | 3 | 1744 | |||

Rbak
Genomic Map