siRNA Details
| Gene Symbol: Rgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301692.1 | NP_001288621.1 | 6 | 737 | ||
| NM_001301694.1 | NP_001288623.1 | 6 | 687 | |||
| NM_021340.4 | NP_067315.1 | 7 | 859 | |||
| GenBank mRNA | AF076930.1 | - | 7 | 779 | ||
| AK044450.1 | - | 7 | 811 | |||
| AK044649.1 | - | 7 | 810 | |||
| AK149272.1 | - | 7 | 810 | |||
| BC082611.1 | - | 7 | 796 | |||
| BC086689.1 | - | 6 | 1273 | |||
| BC119055.1 | - | 7 | 793 | 8733804 | ||
| BC144937.1 | - | 6 | 672 | |||
| CK623457.1 | - | 6 | 618 | |||
More Information
Set Membership: |
|

Rgr
Genomic Map