siRNA Details
| Gene Symbol: Angptl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020581.2 | NP_065606.2 | 1 | 470 | ||
| GenBank mRNA | AB054540.1 | - | 1 | 356 | ||
| AF123261.1 | - | 1 | 401 | |||
| AF169313.1 | - | 1 | 333 | |||
| AF278699.1 | - | 1 | 405 | |||
| AK009161.1 | - | 1 | 176 | |||
| AK014564.1 | - | 471 | ||||
| AK032491.1 | - | 1 | 439 | |||
| AK132761.1 | - | 1 | 439 | |||
| AK134419.1 | - | 1 | 439 | |||
| BC006611.1 | - | 1 | 422 | 3966729 | ||
| BC021343.1 | - | 1 | 436 | 4194173 | ||
| BC025797.1 | - | 1 | 406 | 5137159 | ||
| KJ650202.1 | - | 1 | 287 | |||

Angptl4
Genomic Map