siRNA Details
| Gene Symbol: Cdc42se1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038708.3 | NP_001033797.1 | 3 | 405 | ||
| NM_172395.3 | NP_765983.1 | 3 | 775 | |||
| GenBank mRNA | AK004859.1 | - | 3 | 420 | ||
| AK019052.1 | - | 3 | 417 | |||
| AK028573.1 | - | 3 | 417 | |||
| BC029194.1 | - | 3 | 392 | |||
| BC060964.1 | - | 3 | 397 | 6743495 | ||
| CF105339.1 | - | 3 | 405 | |||

Cdc42se1
Genomic Map