siRNA Details
| Gene Symbol: Rqcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021383.5 | NP_067358.1 | 963 | |||
| GenBank mRNA | AF221849.1 | - | 6 | 585 | ||
| AK005025.2 | - | 6 | 965 | |||
| BC050898.1 | - | 6 | 764 | 6401907 | ||
| BC051948.1 | - | 802 | 5708192 | |||

Rqcd1
Genomic Map