siRNA Details
| Gene Symbol: Cldn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160096.1 | NP_001153568.1 | 4 | 770 | ||
| NM_021386.4 | NP_067361.2 | 4 | 587 | |||
| NM_023878.3 | NP_076367.2 | 4 | 827 | |||
| GenBank mRNA | AF124425.1 | - | 4 | 556 | ||
| AK020131.1 | - | 4 | 791 | |||
| BC012238.1 | - | 2 | 128 | |||
| BC029019.1 | - | 580 | 5356256 | |||
| CK332009.1 | - | 4 | 392 | |||
| CK794879.1 | - | 4 | 587 | |||

Cldn10
Genomic Map