siRNA Details
| Gene Symbol: Vstm2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021387.3 | NP_067362.1 | 4 | 458 | ||
| GenBank mRNA | AB030198.1 | - | 4 | 459 | ||
| AK013845.1 | - | 4 | 446 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vstm2b| Gene Symbol: Vstm2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021387.3 | NP_067362.1 | 4 | 458 | ||
| GenBank mRNA | AB030198.1 | - | 4 | 459 | ||
| AK013845.1 | - | 4 | 446 | |||