siRNA Details
| Gene Symbol: Sh3kbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135727.2 | NP_001129199.1 | 15 | 2000 | ||
| NM_001135728.2 | NP_001129200.1 | 10 | 1002 | |||
| NM_001290661.1 | NP_001277590.1 | 4 | 682 | |||
| NM_021389.6 | NP_067364.2 | 14 | 1565 | |||
| GenBank mRNA | AK004636.1 | - | 14 | 1565 | ||
| AK007283.1 | - | 4 | 668 | |||
| AK028299.1 | - | 10 | 1006 | |||
| AK049182.1 | - | 15 | 2001 | |||
| AK080930.1 | - | 15 | 1865 | |||
| AK142295.1 | - | 4 | 456 | |||
| AK161512.1 | - | 4 | 683 | |||
| AK161735.1 | - | 10 | 1083 | |||
| AK172022.1 | - | 15 | 1981 | |||
| BC034646.1 | - | 14 | 1570 | |||
| BC052863.1 | - | 14 | 1510 | |||

Sh3kbp1
Genomic Map