siRNA Details
| Gene Symbol: Sall1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021390.3 | NP_067365.2 | 3 | 3395 | ||
| GenBank mRNA | AB051409.1 | - | 2 | 3249 | ||
| BC062937.1 | - | 3 | 3395 | 6839137 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sall1| Gene Symbol: Sall1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021390.3 | NP_067365.2 | 3 | 3395 | ||
| GenBank mRNA | AB051409.1 | - | 2 | 3249 | ||
| BC062937.1 | - | 3 | 3395 | 6839137 | ||