siRNA Details
| Gene Symbol: Zbtb32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021397.2 | NP_067372.2 | 5 | 1209 | ||
| GenBank mRNA | AF241232.1 | - | 4 | 1118 | ||
| AK015881.1 | - | 3 | 292 | |||
| BC137687.1 | - | 4 | 1112 | 8860706 | ||
| BC144837.1 | - | 4 | 874 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001109517, XM_006228793, XM_008759188.

Zbtb32
Genomic Map