siRNA Details
| Gene Symbol: Zbtb32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021397.2 | NP_067372.2 | 2 | 316 | ||
| GenBank mRNA | AF241232.1 | - | 1 | 225 | ||
| BC137687.1 | - | 1 | 219 | 8860706 | ||
| BC144837.1 | - | 1 | 219 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb32| Gene Symbol: Zbtb32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021397.2 | NP_067372.2 | 2 | 316 | ||
| GenBank mRNA | AF241232.1 | - | 1 | 225 | ||
| BC137687.1 | - | 1 | 219 | 8860706 | ||
| BC144837.1 | - | 1 | 219 | |||