siRNA Details
| Gene Symbol: Cst10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021405.2 | NP_067380.1 | 2 | 421 | ||
| GenBank mRNA | AB036743.1 | - | 2 | 421 | ||
| BC048364.1 | - | 2 | 337 | 4920219 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cst10| Gene Symbol: Cst10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021405.2 | NP_067380.1 | 2 | 421 | ||
| GenBank mRNA | AB036743.1 | - | 2 | 421 | ||
| BC048364.1 | - | 2 | 337 | 4920219 | ||