siRNA Details
| Gene Symbol: Trem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021406.5 | NP_067381.1 | 2 | 383 | ||
| GenBank mRNA | AF241219.1 | - | 2 | 338 | ||
| AK089439.1 | - | 2 | 352 | |||
| AK171753.1 | - | 2 | 352 | |||
| BY203861.1 | - | 2 | 365 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trem1| Gene Symbol: Trem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021406.5 | NP_067381.1 | 2 | 383 | ||
| GenBank mRNA | AF241219.1 | - | 2 | 338 | ||
| AK089439.1 | - | 2 | 352 | |||
| AK171753.1 | - | 2 | 352 | |||
| BY203861.1 | - | 2 | 365 | |||