siRNA Details
| Gene Symbol: Fam184b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021416.3 | NP_067391.3 | 8 | 1866 | ||
| GenBank mRNA | AB093291.1 | - | 7 | 1395 | ||
| AB257856.1 | - | 8 | 1979 | |||
| AK081567.1 | - | 8 | 1655 | |||
| BC056379.1 | - | 8 | 1673 | 6850901 | ||

Fam184b
Genomic Map