siRNA Details
| Gene Symbol: Fam184b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021416.3 | NP_067391.3 | 1, 2 | 408 | ||
| GenBank mRNA | AB257856.1 | - | 1, 2 | 521 | ||
| AK081567.1 | - | 1, 2 | 192 | |||
| BC056379.1 | - | 1, 2 | 215 | 6850901 | ||
| CF539972.1 | - | 1, 2 | 408 | |||

Fam184b
Genomic Map