siRNA Details
| Gene Symbol: Stk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021420.3 | NP_067395.1 | 3, 4 | 288 | ||
| GenBank mRNA | AF271360.1 | - | 3, 4 | 301 | ||
| AK028838.1 | - | 3, 4 | 290 | |||
| U49950.1 | - | 1, 2 | 148 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stk4| Gene Symbol: Stk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021420.3 | NP_067395.1 | 3, 4 | 288 | ||
| GenBank mRNA | AF271360.1 | - | 3, 4 | 301 | ||
| AK028838.1 | - | 3, 4 | 290 | |||
| U49950.1 | - | 1, 2 | 148 | |||