siRNA Details
| Gene Symbol: Dnaja4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021422.4 | NP_067397.1 | 5, 6 | 1050 | ||
| GenBank mRNA | AB032401.1 | - | 6, 7 | 917 | ||
| AK003903.1 | - | 1, 2 | 191 | |||
| AK016666.1 | - | 4, 5 | 647 | |||
| AK046476.1 | - | 5, 6 | 1051 | |||
| AK076175.1 | - | 5, 6 | 1051 | |||
| AK141909.1 | - | 5, 6 | 1052 | |||
| BC022948.1 | - | 3, 4 | 548 | |||
| BC147484.1 | - | 5, 6 | 898 | 9056231 | ||
| BC147486.1 | - | 5, 6 | 898 | 9056239 | ||

Dnaja4
Genomic Map